MAN424069210 Manchester Organics

(2-Oxo-1H-pyridin-3-yl)boronic acid

  • MDL number MFCD12964572
  • Molecular Weight : 138.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/U45420
Availability details for
Quantity
  • Availability
  • Add to Cart