MAN424069288 Manchester Organics

Quinolin-3-ol

  • MDL number MFCD00169018
  • Molecular Weight : 145.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Z45494
Availability details for
Quantity
  • Availability
  • Add to Cart