MAN424069432 Manchester Organics

1H-1,2,4-Triazole-5-carbonitrile

  • MDL number MFCD11505654
  • Cas Number : 3641-10-9
  • Molecular Weight : 94.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin GB
Product Link https://www.manchesterorganics.com/K45642
Availability details for
Quantity
  • Availability
  • Add to Cart