MAN424069434 Manchester Organics

6-Bromothiazolo[5,4-b]pyridine

  • MDL number MFCD07375013
  • Cas Number : 886372-88-9
  • Molecular Weight : 215.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/M45644
Availability details for
Quantity
  • Availability
  • Add to Cart