MAN424069458 Manchester Organics

4-Bromo-6-chloro-pyridazin-3-amine

  • MDL number MFCD11520891
  • Cas Number : 446273-59-2
  • Molecular Weight : 208.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/N45668
Availability details for
Quantity
  • Availability
  • Add to Cart