MAN424069632 Manchester Organics

tert-Butyl 2,4-dichloro-6,8-dihydro-5H-pyrido[3,4-d]pyrimidine-7-carboxylate

  • MDL number MFCD09025732
  • Cas Number : 916420-27-4
  • Molecular Weight : 304.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 2,4-dichloro-5,8-dihydropyrido[3,4-d]pyrimidine-7(6H)-carboxylate
Inchi Code 1S/C12H15Cl2N3O2/c1-12(2,3)19-11(18)17-5-4-7-8(6-17)15-10(14)16-9(7)13/h4-6H2,1-3H3
Inchi Key SAEOMPAQDWZLHC-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/Z45839
Availability details for
Quantity
  • Availability
  • Add to Cart