MAN424069696 Manchester Organics

Furo[2,3-c]pyridine-2-carboxylic acid

  • MDL number MFCD11869932
  • Molecular Weight : 163.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/F45868
Availability details for
Quantity
  • Availability
  • Add to Cart