MAN424069803 Manchester Organics

tert-Butyl (1S,5R)-6-amino-3-azabicyclo[3.1.0]hexane-3-carboxylate

  • MDL number MFCD09832898
  • Molecular Weight : 198.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B29738
Availability details for
Quantity
  • Availability
  • Add to Cart