MAN424069807 Manchester Organics

(3aR,4S,7R,7aS) 4,7-Methano-1H-isoindole-1,3(2H)-dione

  • MDL number MFCD12964181
  • Molecular Weight : 165.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/C45957
Availability details for
Quantity
  • Availability
  • Add to Cart