MAN424069831 Manchester Organics

(5-Bromo-2-thienyl)methanol

  • MDL number MFCD06202626
  • Cas Number : 79387-71-6
  • Molecular Weight : 193.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/M45966
Availability details for
Quantity
  • Availability
  • Add to Cart