MAN424069840 Manchester Organics

2-(Phenylcarbamoyl)benzoic acid

  • MDL number MFCD00029939
  • Cas Number : 4727-29-1
  • Molecular Weight : 241.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/X45975
Availability details for
Quantity
  • Availability
  • Add to Cart