MAN424069863 Manchester Organics

4-(2-Hydroxyethyl)benzonitrile

  • MDL number MFCD00052461
  • Cas Number : 69395-13-7
  • Molecular Weight : 147.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/G45984
Availability details for
Quantity
  • Availability
  • Add to Cart