MAN424069882 Manchester Organics

1-(4-Chloro-2-pyridyl)-N-methyl-methanamine

  • MDL number MFCD07374905
  • Cas Number : 881205-08-9
  • Molecular Weight : 156.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin GB
Product Link https://www.manchesterorganics.com/D46004
Availability details for
Quantity
  • Availability
  • Add to Cart