MAN424069926 Manchester Organics

2-Bromo-5-ethyl-pyridine

  • MDL number MFCD11869552
  • Cas Number : 19842-08-1
  • Molecular Weight : 186.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin GB
Product Link https://www.manchesterorganics.com/R46015
Availability details for
Quantity
  • Availability
  • Add to Cart