MAN424069980 Manchester Organics

2,6-Dichloro-9-isopropyl-purine

  • MDL number MFCD11047295
  • Molecular Weight : 231.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Z46069
Availability details for
Quantity
  • Availability
  • Add to Cart