MAN424070036 Manchester Organics

3-Aminopyrazine-2-carbonitrile

  • MDL number MFCD02091510
  • Molecular Weight : 120.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A46116
Availability details for
Quantity
  • Availability
  • Add to Cart