MAN424070086 Manchester Organics

4-Bromo-2-methylsulfanyl-pyrimidine

  • MDL number MFCD09878932
  • Cas Number : 959236-97-6
  • Molecular Weight : 205.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/E46166
Availability details for
Quantity
  • Availability
  • Add to Cart