MAN424070311 Manchester Organics

4-Bromopyridine-2,6-diamine

  • MDL number MFCD04115308
  • Molecular Weight : 188.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Z46391
Availability details for
Quantity
  • Availability
  • Add to Cart