MAN424070402 Manchester Organics

2-Chloro-4-methyl-pyrimidin-5-amine

  • MDL number MFCD12923128
  • Molecular Weight : 143.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/Y46482
Availability details for
Quantity
  • Availability
  • Add to Cart