MAN424070403 Manchester Organics

5-Chloro-3-methyl-pyridine-2-carboxylic acid

  • MDL number MFCD07375127
  • Cas Number : 886365-46-4
  • Molecular Weight : 171.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-3-methyl-2-pyridinecarboxylic acid
Inchi Code 1S/C7H6ClNO2/c1-4-2-5(8)3-9-6(4)7(10)11/h2-3H,1H3,(H,10,11)
Inchi Key QQNDJNHLMAFNJI-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/Z46483
Availability details for
Quantity
  • Availability
  • Add to Cart