MAN424070496 Manchester Organics

3-Methoxypyridine-4-carboxylic acid

  • MDL number MFCD07368621
  • Cas Number : 654663-32-8
  • Molecular Weight : 153.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/P46612
Availability details for
Quantity
  • Availability
  • Add to Cart