MAN424070506 Manchester Organics

5-Nitro-1,2-benzoxazole

  • MDL number MFCD06659648
  • Cas Number : 39835-28-4
  • Molecular Weight : 164.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/A46622
Availability details for
Quantity
  • Availability
  • Add to Cart