MAN424070539 Manchester Organics

(3R,5S)-4-Aminoadamantan-1-ol hydrochloride

  • MDL number MFCD11099880
  • Molecular Weight : 203.709

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  
Availability details for
Quantity
  • Availability
  • Add to Cart