MAN424070544 Manchester Organics

2-Bromo-9,10-bis(2-naphthyl)anthracene

  • MDL number MFCD11045064
  • Cas Number : 474688-76-1
  • Molecular Weight : 509.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin GB
Product Link https://www.manchesterorganics.com/S46660
Availability details for
Quantity
  • Availability
  • Add to Cart