MAN424070595 Manchester Organics

6-Chloro-1H-pyrazolo[3,4-d]pyrimidine

  • MDL number MFCD10697157
  • Cas Number : 23002-51-9
  • Molecular Weight : 154.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/X46711
Availability details for
Quantity
  • Availability
  • Add to Cart