MAN424070706 Manchester Organics

Methyl 2-bromoimidazo[1,2-a]pyridine-6-carboxylate

  • MDL number MFCD13195435
  • Cas Number : 1042141-37-6
  • Molecular Weight : 255.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-bromoimidazo[1,2-a]pyridine-6-carboxylate
Inchi Code 1S/C9H7BrN2O2/c1-14-9(13)6-2-3-8-11-7(10)5-12(8)4-6/h2-5H,1H3
Inchi Key QIPNKRNFZTZQBD-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/T46822
Availability details for
Quantity
  • Availability
  • Add to Cart