MAN424070791 Manchester Organics

(3R,4S)-4-Phenylpyrrolidine-3-carboxylic acid hydrochloride

  • MDL number MFCD06659254
  • Molecular Weight : 227.687

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/S46890
Availability details for
Quantity
  • Availability
  • Add to Cart