MAN424070973 Manchester Organics

5-(Trifluoromethyl)pyridin-3-amine

  • MDL number MFCD00128900
  • Molecular Weight : 162.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/G47065
Availability details for
Quantity
  • Availability
  • Add to Cart