MAN424071281 Manchester Organics

3,5-Dichloro-4-methyl benzoic acid

  • MDL number MFCD09743850
  • Molecular Weight : 205.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin GB
Product Link https://www.manchesterorganics.com/C47360
Availability details for
Quantity
  • Availability
  • Add to Cart