MAN424071283 Manchester Organics

7-Fluoro-1H-indole-2-carboxylic acid

  • MDL number MFCD02664429
  • Molecular Weight : 179.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/E47362
Availability details for
Quantity
  • Availability
  • Add to Cart