MAN424071378 Manchester Organics

2-Bromo-3-(trifluoromethoxy)pyridine

  • MDL number MFCD20486158
  • Molecular Weight : 242

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin GB
Product Link https://www.manchesterorganics.com/B12274
Availability details for
Quantity
  • Availability
  • Add to Cart