MAN424071454 Manchester Organics

tert-Butyl 2,4-dioxo-3,8-diazaspiro[4.5]decane-8-carboxylate

  • MDL number MFCD12195867
  • Cas Number : 752234-60-9
  • Molecular Weight : 268.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/W47446
Availability details for
Quantity
  • Availability
  • Add to Cart