MAN424071494 Manchester Organics

3-(Cyanomethyl)benzoic acid

  • MDL number MFCD08234486
  • Cas Number : 5689-33-8
  • Molecular Weight : 161.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(cyanomethyl)benzoic acid
Inchi Code 1S/C9H7NO2/c10-5-4-7-2-1-3-8(6-7)9(11)12/h1-3,6H,4H2,(H,11,12)
Inchi Key DATIHVJZEPOWPT-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/L47483
Availability details for
Quantity
  • Availability
  • Add to Cart