MAN424071663 Manchester Organics

5-Chloro-6-fluoro-1H-indole

  • MDL number MFCD01074495
  • Cas Number : 169674-57-1
  • Molecular Weight : 169.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/U47628
Availability details for
Quantity
  • Availability
  • Add to Cart