MAN424071684 Manchester Organics

(5-Fluoro-2-pyridyl)methanol

  • MDL number MFCD09027049
  • Cas Number : 802325-29-7
  • Molecular Weight : 127.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (5-fluoro-2-pyridinyl)methanol
Inchi Code 1S/C6H6FNO/c7-5-1-2-6(4-9)8-3-5/h1-3,9H,4H2
Inchi Key BKLMFAZXPZQITJ-UHFFFAOYSA-N

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/S47649
Availability details for
Quantity
  • Availability
  • Add to Cart