MAN424071705 Manchester Organics

2-Hydroxyquinoline-7-carboxylic acid

  • MDL number MFCD11557271
  • Molecular Weight : 189.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin GB
Product Link https://www.manchesterorganics.com/Y47724
Availability details for
Quantity
  • Availability
  • Add to Cart