MAN424071710 Manchester Organics

2-[5-Methyl-6-(2-methyl-4-pyridyl)-3-pyridyl]-N-(5-pyrazin-2-yl-2-pyridyl)acetamide

  • MDL number MFCD27501026
  • Cas Number : 1243244-14-5
  • Molecular Weight : 396.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/D47729
Availability details for
Quantity
  • Availability
  • Add to Cart