MAN424071713 Manchester Organics

4-Iodopyridin-2-amine

  • MDL number MFCD09038163
  • Molecular Weight : 220.011

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/G47732
Availability details for
Quantity
  • Availability
  • Add to Cart