MAN424071737 Manchester Organics

6-Bromo-2-fluoro-3-methoxy-pyridine

  • MDL number MFCD04112568
  • Cas Number : 850142-73-3
  • Molecular Weight : 206.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/H47756
Availability details for
Quantity
  • Availability
  • Add to Cart