MAN424071758 Manchester Organics

Sodium-2-[1-[[(1R)-1-[3-[(E)-2-(7-chloro-2-quinolyl)vinyl]phenyl]-3-[2-(1-hydroxy-1-methyl-ethyl)phenyl]propyl]sulfanylmethyl]cyclopropyl]acetate

  • MDL number MFCD00931431
  • Molecular Weight : 608.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/F47777
Availability details for
Quantity
  • Availability
  • Add to Cart