MAN424071764 Manchester Organics

6-Chloro-5-methoxy-pyrimidin-4-amine

  • MDL number MFCD07787635
  • Cas Number : 5018-41-7
  • Molecular Weight : 159.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/M47783
Availability details for
Quantity
  • Availability
  • Add to Cart