MAN424071776 Manchester Organics

(1S,5R)-8-Azabicyclo[3.2.1]octan-3-ol

  • MDL number MFCD00047140
  • Cas Number : 538-09-0
  • Molecular Weight : 127.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin GB
Product Link https://www.manchesterorganics.com/A47795
Availability details for
Quantity
  • Availability
  • Add to Cart