MAN424072055 Manchester Organics

5-Bromo-1-methyl-pyrrole-2-carboxylic acid

  • MDL number MFCD16659627
  • Molecular Weight : 204.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/V48135
Availability details for
Quantity
  • Availability
  • Add to Cart