MAN424072320 Manchester Organics

Methyl 3-chloropyrazine-2-carboxylate

  • MDL number MFCD03093326
  • Molecular Weight : 172.569

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Y48437
Availability details for
Quantity
  • Availability
  • Add to Cart