MAN424072678 Manchester Organics

Ethyl 3-fluoro-1H-pyrrole-2-carboxylate

  • MDL number MFCD11042605
  • Cas Number : 168102-05-4
  • Molecular Weight : 157.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 3-fluoro-1H-pyrrole-2-carboxylate
Inchi Code 1S/C7H8FNO2/c1-2-11-7(10)6-5(8)3-4-9-6/h3-4,9H,2H2,1H3
Inchi Key XKAWEJUJIOUTRS-UHFFFAOYSA-N

Description

Purity 96%
Country of Origin GB
Product Link https://www.manchesterorganics.com/P48751
Availability details for
Quantity
  • Availability
  • Add to Cart