MAN424072728 Manchester Organics

2-(5-Bromo-2-fluoro-phenyl)acetonitrile

  • MDL number MFCD09804785
  • Cas Number : 305800-60-6
  • Molecular Weight : 214.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/U48801
Availability details for
Quantity
  • Availability
  • Add to Cart