MAN424072739 Manchester Organics

(2R)-2-Hydroxypropanamidine hydrochloride

  • MDL number MFCD08752491
  • Molecular Weight : 124.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin GB
Product Link https://www.manchesterorganics.com/F48812
Availability details for
Quantity
  • Availability
  • Add to Cart