MAN424072754 Manchester Organics

6-Chloro-3,4-dihydro-2H-1,4-benzoxazine-8-carboxylic acid

  • MDL number MFCD22384391
  • Cas Number : 134372-47-7
  • Molecular Weight : 213.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/X48827
Availability details for
Quantity
  • Availability
  • Add to Cart