MAN424072794 Manchester Organics

4-Chloro-6-iodopyrimidine

  • MDL number MFCD12756222
  • Molecular Weight : 240.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/N48842
Availability details for
Quantity
  • Availability
  • Add to Cart