MAN424072848 Manchester Organics

2-Bromo-4-chloro-benzonitrile

  • MDL number MFCD00158966
  • Cas Number : 57381-49-4
  • Molecular Weight : 216.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/U48847
Availability details for
Quantity
  • Availability
  • Add to Cart